Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008872.2 | NP_032898.2 | 6 - 7 | 639 | 60 | |
GenBank mRNA | AK135857.1 | - | 6 - 7 | 630 | 60 | |
AK135965.1 | - | 6 - 7 | 631 | 60 | ||
AK159620.1 | - | 6 - 7 | 631 | 60 | ||
AK166639.1 | - | 6 - 7 | 629 | 60 | ||
BC011256.1 | - | 6 - 7 | 636 | 4038404 | 60 | |
BC057967.1 | - | 6 - 7 | 619 | 4979664 | 60 | |
BC061508.1 | - | 6 - 7 | 620 | 5352018 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |