Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slain2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113423.2 | NP_001106894.1 | 1 - 2 | 651 | 74 | |
NM_153567.3 | NP_705795.2 | 1 - 2 | 651 | 74 | ||
GenBank mRNA | AK129365.1 | - | 1 - 2 | 640 | 74 | |
AK146432.1 | - | 1 - 2 | 650 | 74 | ||
BC010836.1 | - | 1 - 2 | 639 | 74 | ||
BC076573.1 | - | 1 - 2 | 584 | 74 | ||
CF728452.1 | - | 2 - 3 | 144 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |