Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map2k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042557.2 | NP_001036022.1 | 12 - 13 | 1477 | 83 | |
NM_001291777.1 | NP_001278706.1 | 11 - 12 | 1429 | 83 | ||
NM_001291778.1 | NP_001278707.1 | 11 - 12 | 1320 | 83 | ||
GenBank mRNA | AB005654.1 | - | 12 - 13 | 1352 | 83 | |
AF060943.1 | - | 11 - 12 | 1323 | 83 | ||
AF060945.1 | - | 11 - 12 | 1343 | 83 | ||
AF060947.1 | - | 12 - 13 | 1391 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Mus musculus Probe spans exons |