Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyrm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175364.4 | NP_780573.1 | 1 - 2 | 71 | 111 | |
GenBank mRNA | AK044791.1 | - | 1 - 2 | 66 | 111 | |
BY131987.1 | - | 1 - 2 | 71 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |