Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rragd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027491.2 | NP_081767.2 | 4 - 5 | 1214 | 74 | |
GenBank mRNA | AK017818.1 | - | 3 - 4 | 574 | 74 | |
BC038137.1 | - | 4 - 5 | 703 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |