Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029465.3 | NP_083741.1 | 3 - 4 | 229 | 94 | |
GenBank mRNA | AK079992.1 | - | 3 - 4 | 230 | 94 | |
BC025069.1 | - | 3 - 4 | 221 | 94 | ||
BC087963.1 | - | 3 - 4 | 229 | 6766377 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |