Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcer2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253737.1 | NP_001240666.1 | 6 - 7 | 413 | 66 | |
NM_001253739.1 | NP_001240668.1 | 6 - 7 | 424 | 66 | ||
NM_001253743.1 | NP_001240672.1 | 7 - 8 | 473 | 66 | ||
NM_001253745.1 | NP_001240674.1 | 5 - 6 | 350 | 66 | ||
NM_001253746.1 | NP_001240675.1 | 5 - 6 | 350 | 66 | ||
NM_001253747.1 | NP_001240676.1 | 6 - 7 | 434 | 66 | ||
NM_013517.4 | NP_038545.1 | 7 - 8 | 554 | 66 | ||
GenBank mRNA | AK156654.1 | - | 7 - 8 | 557 | 66 | |
AK172591.1 | - | 7 - 8 | 557 | 66 | ||
AY069980.1 | - | 5 - 6 | 316 | 66 | ||
AY069981.1 | - | 5 - 6 | 316 | 66 | ||
BC119164.1 | - | 6 - 7 | 421 | 8733913 | 66 | |
BC119190.1 | - | 6 - 7 | 421 | 8733939 | 66 | |
M34163.1 | - | 7 - 8 | 556 | 66 | ||
M99371.1 | - | 7 - 8 | 546 | 66 | ||
S67072.1 | - | 6 - 7 | 405 | 66 | ||
S67073.1 | - | 5 - 6 | 285 | 66 | ||
S67074.1 | - | 6 - 7 | 324 | 66 | ||
X64223.1 | - | 6 - 7 | 413 | 66 | ||
X64224.1 | - | 6 - 7 | 424 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |