Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159573.1 | NP_001153045.1 | 14 - 15 | 1382 | 66 | |
NM_001159574.1 | NP_001153046.1 | 12 - 13 | 1247 | 66 | ||
NM_024183.5 | NP_077145.2 | 13 - 14 | 1274 | 66 | ||
GenBank mRNA | AK005061.1 | - | 13 - 14 | 1243 | 66 | |
AK008561.1 | - | 14 - 15 | 1354 | 66 | ||
AK049672.1 | - | 12 - 13 | 1233 | 66 | ||
AK164105.1 | - | 13 - 14 | 1240 | 66 | ||
AK168464.1 | - | 14 - 15 | 1386 | 66 | ||
BC003263.1 | - | 14 - 15 | 1382 | 3499130 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |