Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038587.4 | NP_001033676.2 | 13 - 14 | 3269 | 95 | |
NM_001146296.1 | NP_001139768.1 | 13 - 14 | 3261 | 95 | ||
NM_019655.3 | NP_062629.3 | 13 - 14 | 3183 | 95 | ||
GenBank mRNA | AF052506.1 | - | 13 - 14 | 3096 | 95 | |
AF291050.1 | - | 13 - 14 | 3090 | 95 | ||
AF291875.1 | - | 12 - 13 | 1658 | 95 | ||
AF291876.1 | - | 13 - 14 | 3168 | 95 | ||
AF291877.1 | - | 12 - 13 | 1736 | 95 | ||
AK076413.1 | - | 13 - 14 | 3220 | 95 | ||
AK089451.1 | - | 13 - 14 | 3232 | 95 | ||
AK147614.1 | - | 13 - 14 | 3232 | 95 | ||
AK156093.1 | - | 12 - 13 | 1750 | 95 | ||
AK172523.1 | - | 12 - 13 | 1750 | 95 | ||
BC042505.1 | - | 13 - 14 | 3117 | 4012994 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |