Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038587.4 | NP_001033676.2 | 5 - 6 | 2032 | 74 | |
NM_001146296.1 | NP_001139768.1 | 5 - 6 | 2024 | 74 | ||
NM_019655.3 | NP_062629.3 | 5 - 6 | 2024 | 74 | ||
GenBank mRNA | AF291875.1 | - | 4 - 5 | 499 | 74 | |
AF291876.1 | - | 5 - 6 | 1931 | 74 | ||
AF291877.1 | - | 4 - 5 | 499 | 74 | ||
AK076413.1 | - | 5 - 6 | 1983 | 74 | ||
AK089451.1 | - | 5 - 6 | 1995 | 74 | ||
AK147614.1 | - | 5 - 6 | 1995 | 74 | ||
AK156093.1 | - | 4 - 5 | 591 | 74 | ||
AK172523.1 | - | 4 - 5 | 591 | 74 | ||
BC042505.1 | - | 5 - 6 | 1958 | 4012994 | 74 | |
CT010323.1 | - | 5 - 6 | 1931 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |