Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rusc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083807.1 | NP_001077276.1 | 7 - 8 | 2021 | 88 | |
NM_001083808.1 | NP_001077277.1 | 6 - 7 | 561 | 88 | ||
NM_028188.2 | NP_082464.2 | 8 - 9 | 2432 | 88 | ||
GenBank mRNA | AK032334.1 | - | 6 - 7 | 1051 | 88 | |
AK039664.1 | - | 8 - 9 | 2360 | 88 | ||
AK042689.1 | - | 7 - 8 | 1969 | 88 | ||
AK170012.1 | - | 6 - 7 | 575 | 88 | ||
AK170210.1 | - | 6 - 7 | 524 | 88 | ||
BC052081.1 | - | 5 - 6 | 755 | 88 | ||
BC056360.1 | - | 7 - 8 | 1959 | 6816267 | 88 | |
BC057034.1 | - | 6 - 7 | 627 | 5687517 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |