Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013604.2 | NP_038632.2 | 3 - 4 | 671 | 71 | |
GenBank mRNA | AK159316.1 | - | 2 - 3 | 358 | 71 | |
AK163982.1 | - | 3 - 4 | 239 | 71 | ||
AK165547.1 | - | 3 - 4 | 246 | 71 | ||
AK165923.1 | - | 3 - 4 | 244 | 71 | ||
AX775987.1 | - | 3 - 4 | 243 | 71 | ||
BB662550.1 | - | 3 - 4 | 263 | 71 | ||
BC023071.1 | - | 3 - 4 | 242 | 5355724 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |