Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc50a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009057.3 | NP_033083.2 | 1 - 2 | 186 | 63 | |
GenBank mRNA | AK014305.1 | - | 1 - 2 | 139 | 63 | |
AK079716.1 | - | 1 - 2 | 140 | 63 | ||
AK170162.1 | - | 1 - 2 | 137 | 63 | ||
BC014292.1 | - | 1 - 2 | 170 | 4190124 | 63 | |
X96618.1 | - | 1 - 2 | 177 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |