Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gon4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242372.1 | NP_001229301.1 | 5564 | 88 | ||
NM_027389.3 | NP_081665.2 | 5513 | 88 | |||
GenBank mRNA | AK122531.1 | - | 4885 | 88 | ||
AK147575.1 | - | 5515 | 88 | |||
AK166365.1 | - | 535 | 88 | |||
BC016616.1 | - | 885 | 88 | |||
BC108377.1 | - | 25 - 26 | 4804 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |