Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163432.1 | NP_001156904.1 | 7 - 8 | 1131 | 72 | |
NM_007712.3 | NP_031738.2 | 7 - 8 | 1134 | 72 | ||
GenBank mRNA | AF033564.1 | - | 7 - 8 | 856 | 72 | |
AK134849.1 | - | 1 - 2 | 1375 | 72 | ||
AK148377.1 | - | 5 - 6 | 2162 | 72 | ||
AK161259.1 | - | 3 - 4 | 1306 | 72 | ||
BC015080.1 | - | 7 - 8 | 1119 | 3995512 | 72 | |
CN538691.1 | - | 2 - 3 | 296 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |