Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025327.2 | NP_079603.1 | 1 - 2 | 78 | 74 | |
GenBank mRNA | AK002479.1 | - | 1 - 2 | 79 | 74 | |
AK003310.1 | - | 1 - 2 | 46 | 74 | ||
BC043030.1 | - | 1 - 2 | 550 | 5693076 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |