Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900005857.mm.1 |
NM_001130525.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005857.mm.1 |
NM_001130525.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005857.mm.1 |
NM_001130526.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005857.mm.1 |
NM_001130526.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005857.mm.1 |
NM_145503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005857.mm.1 |
NM_145503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005857.mm.1 |
TC1900000662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005855.mm.1 |
NM_001130526.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005855.mm.1 |
NM_001130526.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005855.mm.1 |
TC1900000662.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900005858.mm.1 |
NM_001130525.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005858.mm.1 |
NM_001130525.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005858.mm.1 |
NM_001130526.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005858.mm.1 |
NM_001130526.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005858.mm.1 |
NM_145503.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005858.mm.1 |
NM_145503.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900005858.mm.1 |
TC1900000662.mm.1 |
MTA TC |