Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016771.3 | NP_058051.3 | 4 - 5 | 591 | 78 | |
GenBank mRNA | AF026073.1 | - | 4 - 5 | 469 | 78 | |
AK133530.1 | - | 4 - 5 | 510 | 78 | ||
AK160466.1 | - | 4 - 5 | 576 | 78 | ||
BC010752.1 | - | 4 - 5 | 564 | 78 | ||
BC066190.1 | - | 4 - 5 | 544 | 30472198 | 78 | |
U32371.1 | - | 4 - 5 | 541 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |