Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023135.2 | NP_075624.2 | 1 - 2 | 117 | 110 | |
GenBank mRNA | AK015750.1 | - | 1 - 2 | 101 | 110 | |
BB590464.1 | - | 1 - 2 | 118 | 110 | ||
BC034891.1 | - | 1 - 2 | 58 | 1479883 | 110 | |
S78182.1 | - | 1 - 2 | 100 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |