Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019878.4 | NP_063931.1 | 2 - 3 | 248 | 131 | |
GenBank mRNA | AF022894.1 | - | 2 - 3 | 267 | 131 | |
AK033498.1 | - | 2 - 3 | 251 | 131 | ||
AK087595.1 | - | 2 - 3 | 284 | 131 | ||
BC024361.1 | - | 1 - 2 | 207 | 4990345 | 131 | |
U92076.1 | - | 2 - 3 | 193 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |