Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sult1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019878.4 | NP_063931.1 | 4 - 5 | 475 | 136 | |
GenBank mRNA | AF022894.1 | - | 4 - 5 | 494 | 136 | |
AK033498.1 | - | 4 - 5 | 478 | 136 | ||
AK087595.1 | - | 4 - 5 | 511 | 136 | ||
BC024361.1 | - | 3 - 4 | 434 | 4990345 | 136 | |
U92076.1 | - | 4 - 5 | 420 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |