Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133761.3 | NP_598522.3 | 3 - 4 | 419 | 97 | |
GenBank mRNA | AJ344447.1 | - | 3 - 4 | 419 | 97 | |
AK171707.1 | - | 3 - 4 | 343 | 97 | ||
BC066173.1 | - | 3 - 4 | 372 | 30049516 | 97 | |
DV651307.1 | - | 3 - 4 | 449 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |