Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008406.3 | NP_032432.2 | 10 - 11 | 1245 | 87 | |
GenBank mRNA | AK149500.1 | - | 10 - 11 | 1246 | 87 | |
BC013465.1 | - | 10 - 11 | 1229 | 87 | ||
BC028814.1 | - | 10 - 11 | 1245 | 5099284 | 87 | |
X70391.1 | - | 10 - 11 | 1225 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |