Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB073078.1 | - | 3 - 4 | 403 | 69 | |
AB073079.1 | - | 3 - 4 | 404 | 69 | ||
AB073080.1 | - | 3 - 4 | 403 | 69 | ||
AK050922.1 | - | 4 - 5 | 504 | 69 | ||
AK078113.1 | - | 2 - 3 | 295 | 69 | ||
AK142379.1 | - | 2 - 3 | 314 | 69 | ||
BB639043.1 | - | 2 - 3 | 340 | 69 | ||
BC145176.1 | - | 2 - 3 | 294 | 9053727 | 69 | |
BC145850.1 | - | 2 - 3 | 294 | 40131153 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |