Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB073079.1 | - | 6 - 7 | 839 | 89 | |
AB073080.1 | - | 6 - 7 | 838 | 89 | ||
AK078113.1 | - | 5 - 6 | 730 | 89 | ||
AK142379.1 | - | 5 - 6 | 749 | 89 | ||
BC145850.1 | - | 5 - 6 | 729 | 40131153 | 89 | |
S66169.1 | - | 2 - 3 | 151 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |