Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800002623.mm.1 |
NM_153592.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002623.mm.1 |
NM_153592.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002623.mm.1 |
TC0800000345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002624.mm.1 |
NM_153592.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002624.mm.1 |
NM_153592.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002624.mm.1 |
TC0800000345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002625.mm.1 |
NM_153592.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002625.mm.1 |
NM_153592.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002625.mm.1 |
TC0800000345.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800002626.mm.1 |
NM_153592.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002626.mm.1 |
NM_153592.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800002626.mm.1 |
TC0800000345.mm.1 |
MTA TC |