Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153592.2 | NP_705820.1 | 4 - 5 | 357 | 99 | |
GenBank mRNA | AK030557.1 | - | 4 - 5 | 301 | 99 | |
BC036333.1 | - | 4 - 5 | 322 | 3590904 | 99 | |
BC038374.1 | - | 4 - 5 | 297 | 5355446 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |