Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006122.1 | - | 3 - 4 | 325 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |