Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dido1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291433.1 | NP_001278362.1 | 8 - 9 | 2536 | 91 | |
NM_175551.4 | NP_780760.2 | 8 - 9 | 2536 | 91 | ||
NM_177852.4 | NP_808520.2 | 8 - 9 | 2484 | 91 | ||
GenBank mRNA | AK049707.1 | - | 2 - 3 | 1151 | 91 | |
AK129117.1 | - | 8 - 9 | 2484 | 91 | ||
AY425951.1 | - | 8 - 9 | 2438 | 91 | ||
AY425952.1 | - | 8 - 9 | 2438 | 91 | ||
BC044755.1 | - | 2 - 3 | 635 | 91 | ||
BC138712.1 | - | 7 - 8 | 2313 | 8861734 | 91 | |
BC138713.1 | - | 7 - 8 | 2313 | 8861735 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |