Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177706.1 | NP_001171177.1 | 4 - 5 | 860 | 79 | |
NM_001177707.1 | NP_001171178.1 | 5 - 6 | 731 | 79 | ||
NM_001177708.1 | NP_001171179.1 | 4 - 5 | 450 | 79 | ||
NM_001177709.1 | NP_001171180.1 | 4 - 5 | 450 | 79 | ||
NM_001177710.1 | NP_001171181.1 | 3 - 4 | 340 | 79 | ||
NM_145760.3 | NP_665703.2 | 5 - 6 | 663 | 79 | ||
GenBank mRNA | AK030295.1 | - | 5 - 6 | 619 | 79 | |
AK030520.1 | - | 4 - 5 | 862 | 79 | ||
AK134402.1 | - | 5 - 6 | 673 | 79 | ||
AK145879.1 | - | 5 - 6 | 647 | 79 | ||
AK168007.1 | - | 5 - 6 | 644 | 79 | ||
AK168602.1 | - | 4 - 5 | 431 | 79 | ||
BC016228.1 | - | 3 - 4 | 298 | 79 | ||
BC027794.1 | - | 5 - 6 | 670 | 79 | ||
BC052922.1 | - | 4 - 5 | 398 | 6312351 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |