Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083328.1 | NP_001076797.1 | 3 - 4 | 211 | 84 | |
GenBank mRNA | AK040689.1 | - | 3 - 4 | 213 | 84 | |
BF012021.1 | - | 2 - 3 | 135 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus 1700 Probe spans exons |