Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045541.1 | 3 - 4 | 505 | 97 | ||
GenBank mRNA | AF043276.1 | - | 3 - 4 | 556 | 97 | |
AK043275.1 | - | 3 - 4 | 455 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |