Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB073079.1 | - | 7 - 8 | 912 | 61 | |
AK078113.1 | - | 6 - 7 | 803 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |