Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007834.3 | NP_031860.1 | 5 - 6 | 741 | 88 | |
GenBank mRNA | AB001990.1 | - | 5 - 6 | 556 | 88 | |
AK077485.1 | - | 5 - 6 | 555 | 88 | ||
AK077628.1 | - | 5 - 6 | 547 | 88 | ||
BC003740.1 | - | 5 - 6 | 529 | 3592608 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |