Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111026.2 | NP_001104496.1 | 2 - 3 | 562 | 90 | |
NM_001111027.2 | NP_001104497.1 | 3 - 4 | 756 | 90 | ||
NM_009822.3 | NP_033952.1 | 2 - 3 | 494 | 90 | ||
GenBank mRNA | AB221624.1 | - | 1 - 2 | 141 | 90 | |
AK032132.1 | - | 2 - 3 | 429 | 90 | ||
AK049615.1 | - | 2 - 3 | 300 | 90 | ||
AK142006.1 | - | 2 - 3 | 563 | 90 | ||
BC139200.1 | - | 2 - 3 | 220 | 8862222 | 90 | |
BC139201.1 | - | 2 - 3 | 224 | 8862223 | 90 | |
BY125464.1 | - | 2 - 3 | 227 | 90 | ||
D32007.1 | - | 2 - 3 | 494 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |