Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144517.4 | NP_653100.1 | 13 - 14 | 1090 | 95 | |
GenBank mRNA | AK012836.1 | - | 13 - 14 | 1041 | 95 | |
AK076128.1 | - | 13 - 14 | 1038 | 95 | ||
AK083084.1 | - | 13 - 14 | 1061 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |