Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002011.3 | NP_001002011.2 | 1 - 2 | 603 | 69 | |
NM_001111102.2 | NP_001104572.1 | 1 - 2 | 603 | 69 | ||
GenBank mRNA | AK004619.2 | - | 1 - 2 | 558 | 69 | |
AK147150.1 | - | 1 - 2 | 559 | 69 | ||
AK149998.1 | - | 1 - 2 | 559 | 69 | ||
AK150391.1 | - | 1 - 2 | 559 | 69 | ||
AK150501.1 | - | 1 - 2 | 559 | 69 | ||
AK150624.1 | - | 1 - 2 | 559 | 69 | ||
AK152539.1 | - | 1 - 2 | 559 | 69 | ||
AK152646.1 | - | 1 - 2 | 559 | 69 | ||
AK152846.1 | - | 1 - 2 | 559 | 69 | ||
AK153295.1 | - | 1 - 2 | 501 | 69 | ||
AK161221.1 | - | 1 - 2 | 424 | 69 | ||
AK180919.1 | - | 1 - 2 | 54 | 69 | ||
BC015302.1 | - | 1 - 2 | 479 | 4240057 | 69 | |
BC094020.1 | - | 1 - 2 | 465 | 4187374 | 69 | |
BQ715532.1 | - | 1 - 2 | 606 | 69 | ||
DQ832702.1 | - | 1 - 2 | 354 | 69 | ||
DQ832703.1 | - | 1 - 2 | 354 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |