Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lmna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002011.3 | NP_001002011.2 | 4 - 5 | 1057 | 96 | |
NM_001111102.2 | NP_001104572.1 | 4 - 5 | 1057 | 96 | ||
NM_019390.3 | NP_062263.1 | 4 - 5 | 528 | 96 | ||
GenBank mRNA | AK004619.2 | - | 4 - 5 | 1012 | 96 | |
AK147150.1 | - | 4 - 5 | 1013 | 96 | ||
AK149998.1 | - | 4 - 5 | 1013 | 96 | ||
AK150391.1 | - | 4 - 5 | 1013 | 96 | ||
AK150501.1 | - | 4 - 5 | 1013 | 96 | ||
AK150624.1 | - | 4 - 5 | 1013 | 96 | ||
AK152539.1 | - | 4 - 5 | 1013 | 96 | ||
AK152646.1 | - | 4 - 5 | 1013 | 96 | ||
AK152846.1 | - | 4 - 5 | 1013 | 96 | ||
AK161221.1 | - | 4 - 5 | 878 | 96 | ||
AK167858.1 | - | 4 - 5 | 1010 | 96 | ||
AK215823.1 | - | 1 - 2 | 136 | 96 | ||
BC015302.1 | - | 4 - 5 | 933 | 4240057 | 96 | |
BC094020.1 | - | 4 - 5 | 919 | 4187374 | 96 | |
D13181.1 | - | 1 - 2 | 106 | 96 | ||
D14850.1 | - | 4 - 5 | 528 | 96 | ||
DQ832702.1 | - | 4 - 5 | 808 | 96 | ||
DQ832703.1 | - | 4 - 5 | 808 | 96 | ||
X14170.1 | - | 4 - 5 | 1002 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |