Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghitm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199122.1 | NP_001186051.1 | 2 - 3 | 349 | 66 | |
NM_078478.5 | NP_510963.1 | 2 - 3 | 302 | 66 | ||
GenBank mRNA | AF412297.1 | - | 2 - 3 | 246 | 66 | |
AK005636.1 | - | 2 - 3 | 246 | 66 | ||
AK093887.1 | - | 2 - 3 | 278 | 66 | ||
AK134592.1 | - | 2 - 3 | 232 | 66 | ||
AK146433.1 | - | 2 - 3 | 209 | 66 | ||
AK146853.1 | - | 2 - 3 | 265 | 66 | ||
AK151488.1 | - | 2 - 3 | 279 | 66 | ||
AK158904.1 | - | 2 - 3 | 292 | 66 | ||
AK159695.1 | - | 2 - 3 | 276 | 66 | ||
AK159724.1 | - | 2 - 3 | 276 | 66 | ||
AK164122.1 | - | 2 - 3 | 247 | 66 | ||
AK167970.1 | - | 2 - 3 | 273 | 66 | ||
AK168022.1 | - | 2 - 3 | 279 | 66 | ||
AK168678.1 | - | 2 - 3 | 350 | 66 | ||
AK168800.1 | - | 2 - 3 | 258 | 66 | ||
BC008622.1 | - | 2 - 3 | 280 | 3588758 | 66 | |
BC010224.1 | - | 2 - 3 | 282 | 3500685 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |