Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127355.1 | NP_001120827.1 | 5 - 6 | 754 | 100 | |
NM_001127356.1 | NP_001120828.1 | 4 - 5 | 976 | 100 | ||
NM_010122.2 | NP_034252.2 | 5 - 6 | 609 | 100 | ||
GenBank mRNA | AK154364.1 | - | 5 - 6 | 517 | 100 | |
AK158282.1 | - | 5 - 6 | 518 | 100 | ||
BC021884.1 | - | 5 - 6 | 496 | 4168186 | 100 | |
M98035.1 | - | 4 - 5 | 596 | 100 | ||
M98036.1 | - | 5 - 6 | 562 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |