Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127355.1 | NP_001120827.1 | 6 - 7 | 857 | 58 | |
NM_001127356.1 | NP_001120828.1 | 5 - 6 | 1079 | 58 | ||
NM_010122.2 | NP_034252.2 | 6 - 7 | 712 | 58 | ||
GenBank mRNA | AK154364.1 | - | 6 - 7 | 620 | 58 | |
AK158282.1 | - | 6 - 7 | 621 | 58 | ||
BC021884.1 | - | 6 - 7 | 599 | 4168186 | 58 | |
M98035.1 | - | 5 - 6 | 699 | 58 | ||
M98036.1 | - | 6 - 7 | 665 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |