Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018878.3 | NP_061366.2 | 5 - 6 | 708 | 113 | |
GenBank mRNA | AF104261.1 | - | 5 - 6 | 708 | 113 | |
AK144606.1 | - | 5 - 6 | 570 | 113 | ||
AK148141.1 | - | 5 - 6 | 577 | 113 | ||
BC066014.1 | - | 5 - 6 | 497 | 5704566 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |