Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201413.1 | NP_001188342.1 | 12 - 13 | 2210 | 59 | |
NM_001201414.1 | NP_001188343.1 | 11 - 12 | 2213 | 59 | ||
NM_001201415.1 | NP_001188344.1 | 11 - 12 | 2147 | 59 | ||
NM_001201416.1 | NP_001188345.1 | 2 - 3 | 313 | 59 | ||
NM_009686.2 | NP_033816.1 | 13 - 14 | 2216 | 59 | ||
GenBank mRNA | AK004792.1 | - | 13 - 14 | 2070 | 59 | |
AK153260.1 | - | 11 - 12 | 2179 | 59 | ||
AK167649.1 | - | 12 - 13 | 2075 | 59 | ||
AK169959.1 | - | 11 - 12 | 2088 | 59 | ||
AK172405.1 | - | 2 - 3 | 317 | 59 | ||
BC042797.1 | - | 3 - 4 | 355 | 59 | ||
BC076587.1 | - | 11 - 12 | 1993 | 30550087 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |