Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028234.1 | NP_082510.1 | 7 - 8 | 1030 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |