Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fip1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159573.1 | NP_001153045.1 | 8 - 9 | 832 | 94 | |
NM_001159574.1 | NP_001153046.1 | 8 - 9 | 832 | 94 | ||
NM_024183.5 | NP_077145.2 | 8 - 9 | 832 | 94 | ||
GenBank mRNA | AK005061.1 | - | 8 - 9 | 801 | 94 | |
AK008561.1 | - | 8 - 9 | 804 | 94 | ||
AK049672.1 | - | 8 - 9 | 818 | 94 | ||
AK164105.1 | - | 8 - 9 | 798 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |