Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172689.3 | NP_766277.3 | 13 - 14 | 2049 | 112 | |
GenBank mRNA | AK049305.1 | - | 4 - 5 | 640 | 112 | |
AK052595.1 | - | 9 - 10 | 1303 | 112 | ||
AK128929.1 | - | 13 - 14 | 1984 | 112 | ||
AK155930.1 | - | 13 - 14 | 1985 | 112 | ||
AY553221.1 | - | 13 - 14 | 1925 | 112 | ||
BC027369.1 | - | 13 - 14 | 2028 | 112 | ||
BC106095.1 | - | 13 - 14 | 1969 | 112 | ||
BC127163.1 | - | 13 - 14 | 1915 | 112 | ||
BC152540.1 | - | 13 - 14 | 1917 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |