Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253737.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253737.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253739.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253739.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253743.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253743.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253745.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253745.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253746.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253746.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253747.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_001253747.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_013517.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
NM_013517.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800013286.mm.1 |
TC0800001616.mm.1 |
MTA TC |