Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039578.3 | NP_001034667.3 | 2 - 3 | 379 | 70 | |
GenBank mRNA | AK045240.1 | - | 2 - 3 | 380 | 70 | |
AK087482.1 | - | 2 - 3 | 380 | 70 | ||
AK193949.1 | - | 2 - 3 | 69 | |||
BC042653.1 | - | 2 - 3 | 359 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |