Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrnad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153562.4 | NP_705790.3 | 3 - 4 | 663 | 67 | |
GenBank mRNA | AK030684.1 | - | 2 - 3 | 783 | 67 | |
AK035389.1 | - | 3 - 4 | 615 | 67 | ||
AK082223.1 | - | 3 - 4 | 596 | 67 | ||
AK172049.1 | - | 2 - 3 | 768 | 67 | ||
BC012224.1 | - | 3 - 4 | 555 | 67 | ||
BC023814.1 | - | 2 - 3 | 503 | 5353932 | 67 | |
BC094052.1 | - | 2 - 3 | 1187 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |