Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010684.2 | NP_034814.2 | 3 - 4 | 634 | 151 | |
GenBank mRNA | AK004637.1 | - | 3 - 4 | 501 | 151 | |
AK159096.1 | - | 3 - 4 | 500 | 151 | ||
AK172007.1 | - | 3 - 4 | 516 | 151 | ||
AY069968.1 | - | 3 - 4 | 571 | 151 | ||
BC006044.1 | - | 1 - 2 | 176 | 151 | ||
BC049097.1 | - | 3 - 4 | 608 | 5716524 | 151 | |
J03881.1 | - | 2 - 3 | 308 | 151 | ||
M25244.1 | - | 3 - 4 | 489 | 151 | ||
M32015.1 | - | 3 - 4 | 458 | 151 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |